missing translation for 'onlineSavingsMsg'
Learn More

JG Finneran Associates™ 8-425 mm Thread Top Glass Vial

Product Code. 12397059
Click to view available options
Color:
Amber
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12397059

Brand: JG Finneran Associates™ 32008E1232A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with autosamplers that require a narrow opening.

  • Narrow opening
  • Designed to fit in a variety of autosamplers

Specifications

Color Amber
Material Borosilicate Glass
Volume (Metric) 2 mL
No. per Pack 100
Quantity 100 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.