missing translation for 'onlineSavingsMsg'
Learn More

8-Aminooctanoic acid, 99%

Product Code. 11463597
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463597 25 g 25g
11443597 1 g 1g
11453597 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463597 Supplier Thermo Scientific Alfa Aesar Supplier No. A17353.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1002-57-9 | C8H17NO2 | 159.23 g/mol

8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Aminooctanoic acid is involved in the preparation of lactams using enzyme catalyzed aminolysis.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-57-9
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00008245
InChI Key UQXNEWQGGVUVQA-UHFFFAOYSA-N
Synonym 8-aminocaprylic acid, octanoic acid, 8-amino, omega-aminocaprylic acid, 8-amino caprylic acid, 8-amino-octanoic acid, .omega.-aminocaprylic acid, 8-amino capylic acid, 8ac, 8-aminocapylic acid, pubchem13165
PubChem CID 66085
IUPAC Name 8-aminooctanoic acid
SMILES NCCCCCCCC(O)=O

Specifications

Melting Point 193°C to 196°C
Quantity 25 g
Solubility Information Soluble aq. soln..
Formula Weight 159.23
Percent Purity 99%
Chemical Name or Material 8-Aminooctanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.