missing translation for 'onlineSavingsMsg'
Learn More

8-Aminoquinoline, 98+%

Product Code. 10051270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10051270 5 g 5g
10346810 10 g 10g
10125543 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10051270 Supplier Thermo Scientific Acros Supplier No. 123710050
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 578-66-5 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-66-5
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00006809
InChI Key WREVVZMUNPAPOV-UHFFFAOYSA-N
Synonym 8-aminoquinoline, 8-quinolinamine, 8-quinolylamine, quinoline, 8-amino, quinolin-8-ylamine, unii-u34eav21tg, 8-amino quinoline, ccris 1683, u34eav21tg, 8-amino-quinoline
PubChem CID 11359
IUPAC Name quinolin-8-amine
SMILES C1=CC2=C(C(=C1)N)N=CC=C2

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 62.0°C to 65.0°C
Color Beige-Brown to Green
Boiling Point 174.0°C (26.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 22, 450
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol, insoluble in ether
Formula Weight 144.18
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 8-Aminoquinoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.