missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 8-Azaguanine

Product Code. 16488766
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488766 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488766 Supplier MedChemExpress Supplier No. HYB1468
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 8-Azaguanine is a purine analogue that shows antineoplastic activity. 8-Azaguanine functions as an antimetabolite and easily incorporates into ribonucleic acids, interfering with normal biosynthetic pathways, thus inhibiting cellular growth.

Specifications

Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 152.11 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 134-58-7
Product Type Natural Products
Purity 98.0%
Solubility DMSO : 2.44 mg/mL (16.04 mM∣Need ultrasonic)
Target Nucleoside Antimetabolite/Analog∣Endogenous Metabolite
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.