missing translation for 'onlineSavingsMsg'
Learn More

8-Azaguanine (10 mM in DMSO), TRC

Product Code. 30392639
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392639 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392639 Supplier Toronto Research Chemicals Supplier No. TRCKIT41631X1ML
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material 8-Azaguanine
CAS 134-58-7
Molecular Formula C4 H4 N6 O
Quantity 1x1 mL
Synonym 5-Amino-3,6-dihydro-7H-1,2,3-triazolo(4,5-d)pyrimidin-7-one, Azaguanine, Azan, Guanazol, Guanazolo, Pathocidin
InChI Formula InChI=1S/C4H4N6O/c5-4-6-2-1(3(11)7-4)8-10-9-2/h(H4,5,6,7,8,9,10,11)
SMILES NC1=Nc2nn[nH]c2C(=O)N1
IUPAC Name 5-amino-1,6-dihydrotriazolo[4,5-d]pyrimidin-7-one
Molecular Weight (g/mol) 152.11
Formula Weight 152.0447
Concentration or Composition (by Analyte or Components) 10 mM
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.