missing translation for 'onlineSavingsMsg'
Learn More

8-Azaguanine 98%

Product Code. 15801012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15801012 5 g 5g
15891002 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15801012 Supplier Thermo Scientific Acros Supplier No. 229920050
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 134-58-7 | C4H4N6O | 152.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-58-7
Molecular Formula C4H4N6O
Molecular Weight (g/mol) 152.12
MDL Number MFCD00056937
InChI Key LPXQRXLUHJKZIE-UHFFFAOYSA-N
Synonym 8-azaguanine, guanazolo, azaguanine-8, azaguanine, guanazol, pathocidin, pathocidine, triazologuanine, azan, 8 ag
PubChem CID 8646
ChEBI CHEBI:63486
SMILES NC1=NC(=O)C2=NNNC2=N1

Specifications

Color Yellow
Infrared Spectrum Authentic
Quantity 5 g
Merck Index 15, 889
IUPAC Name 5-amino-2,3-dihydrotriazolo[4,5-d]pyrimidin-7-one
Formula Weight 152.12
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 8-Azaguanine, 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.