missing translation for 'onlineSavingsMsg'
Learn More

8-benzyl-8-azabicyclo[3.2.1]octan-3-endo-ol, 95%, Thermo Scientific™

Product Code. 11386845
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11386845 5 g Amber Glass Bottle 5g
10787865 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11386845 Supplier Thermo Scientific Maybridge Supplier No. CC73715DE
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 18717-73-2
Molecular Formula C14H19NO
Molecular Weight (g/mol) 217.312
MDL Number MFCD09966164
InChI Key HCBGIBWAPOFRKI-PBWFPOADSA-N
Synonym n-benzylnortropine, 1r,5s-8-benzyl-8-azabicyclo 3.2.1 octan-3-ol, endo-8-benzyl-8-azabicyclo 3.2.1 octan-3-ol, 1s,5r-8-benzyl-8-azabicyclo 3.2.1 octan-3-ol, 1r,3r,5s-8-benzyl-8-azabicyclo 3.2.1 octan-3-ol, 1r,3s,5s-8-benzyl-8-azabicyclo 3.2.1 octan-3-ol
PubChem CID 14762744
IUPAC Name (1R,5S)-8-benzyl-8-azabicyclo[3.2.1]octan-3-ol
SMILES C1CC2CC(CC1N2CC3=CC=CC=C3)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 217.31
Percent Purity 95%
Chemical Name or Material 8-Benzyl-8-azabicyclo[3.2.1]octan-3-endo-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.