missing translation for 'onlineSavingsMsg'
Learn More

8-Bromoisoquinoline, 97%

Product Code. 15599573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15599573 1 g 1g
15509583 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15599573 Supplier Thermo Scientific Acros Supplier No. 458760010
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 63927-22-0 | C9H6BrN | 208.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63927-22-0
Molecular Formula C9H6BrN
Molecular Weight (g/mol) 208.06
MDL Number MFCD04973298
InChI Key DPRIHFQFWWCIGY-UHFFFAOYSA-N
Synonym 8-bromo-isoquinoline, isoquinoline,8-bromo, isoquinoline, 8-bromo, d-lactosyl fluoride, pubchem6213, acmc-1b6s3, ksc352q0f, abbypharma ap-12-10879
PubChem CID 9859134
IUPAC Name 8-bromoisoquinoline
SMILES BrC1=C2C=NC=CC2=CC=C1

Specifications

Melting Point 78.0°C to 81.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 208.06
Percent Purity 97%
Chemical Name or Material 8-Bromoisoquinoline
compliance-icons
Product Identifier
  • 8-Bromoisoquinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.