missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ 3500 Dx Genetic Analyzer Capillary Array, 50 cm

Product Code. 15938826 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Array
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15938826 1 Array Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15938826 Supplier Applied Biosystems™ Supplier No. 4404684
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A radio-frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.

These internally uncoated 50-cm capillaries are supplied in assemblies of 8 capillaries per array, with a built-in frame for easy installation. A radio-frequency identification (RFID) tag is built into the frame for tracking critical run information, which is then reported on the software dashboard. The array can be used for 160 injections in Dx mode.

This product is intended for in vitro diagnostic use on the 3500 Dx Genetic Analyzer and is not validated for use with other systems or applications. Refer to the 3500 Dx Genetic Analyzer User Guide for further details.

These internally uncoated 50 cm capillaries are supplied in preassembled set assemblies of 8 capillaries per array, with a built-in frame for easy installation.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Capillary Length 50 cm
Content And Storage Ambient, 1 array with 8 capillaries
For Use With (Equipment) 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Product Type 3500 Dx Genetic Analyzer
Quantity 1 Array
Shipping Condition Room Temperature

For In Vitro Diagnostic Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.