missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ 8-Channel Filter Plates

For use with Agilent 850-DS dissolution sampling station

Brand:  Cytiva 7707-3000

Code : NEW

Additional Details : Weight : 1.80000kg

 View more versions of this product

Product Code. 15215928

  • 680.00 € / Pack of 50
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • 0.45μm membrane pore size
  • Available in PTFE, Nylon, PES, and GMF filter media
  • Disposable

Automated dissolution testing

Specifications

Specifications

Agilent 850-DS Dissolution Sampling Station
PTFE
0.45 μm
Automated dissolution testing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers