missing translation for 'onlineSavingsMsg'
Learn More

(±)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine Hydrochloride, TRC

Product Code. 30360631
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360631 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360631 Supplier Toronto Research Chemicals Supplier No. TRCC602023250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1431697-94-7
Molecular Formula C11H14ClN . HCl
Quantity 250 mg
Synonym rac-8-Chloro-2,3,4,5-tetrahydro-1-methyl-1H-3-benzazepine Hydrochloride rac-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine Hydrochloride
InChI Formula InChI=1S/C11H14ClN.ClH/c1-8-7-13-5-4-9-2-3-10(12)6-11(8)9;/h2-3,6,8,13H,4-5,7H2,1H3;1H
SMILES O.Cl.C[C@H]1CNCCc2ccc(Cl)cc12
IUPAC Name 7-chloro-5-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine;hydrochloride
Molecular Weight (g/mol) 232.15
Formula Weight 249.0687
Chemical Name or Material (±)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.