missing translation for 'onlineSavingsMsg'
Learn More

8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine, 97%, Thermo Scientific™

Product Code. 10692823
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10692823 250 mg 250mg
10563042 1 g 1g
10002464 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10692823 Supplier Thermo Scientific Maybridge Supplier No. CD08167CB
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 131728-94-4
Molecular Formula C9H8ClFO2
Molecular Weight (g/mol) 202.61
MDL Number MFCD00052603
InChI Key FMONGDHUPLQOCP-UHFFFAOYSA-N
Synonym 8-chloromethyl-6-fluoro-4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-4h-benzo d 1,3 dioxine, 4h-1,3-benzodioxin,8-chloromethyl-6-fluoro, 8-chloromethyl-6-fluoro-2,4-dihydro-1,3-benzodioxine, 8-chloromethyl-6-fluorobenzo-1,3-dioxane, acmc-1c5x1, 8-chloromethyl-6-fluoro-4h-benzo 1,3 dioxine, 8-chloromethyl-6-fluoro-2h,4h-1,3-benzodioxine, 8-chloromethyl-6-fluoro-2h,4h-benzo e 1,3-dioxin
PubChem CID 2780137
IUPAC Name 8-(chloromethyl)-6-fluoro-4H-1,3-benzodioxine
SMILES FC1=CC(CCl)=C2OCOCC2=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 202.61
Percent Purity 97%
Chemical Name or Material 8-(Chloromethyl)-6-fluoro-4H-1, 3-benzodioxine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.