missing translation for 'onlineSavingsMsg'
Learn More

8-Dehydrocholesterol, TRC

Product Code. 30363974
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30363974 50 mg 50mg
30356756 25 mg 25mg
30352522 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363974 Supplier Toronto Research Chemicals Supplier No. TRCD23029550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70741-38-7
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.64
Synonym Cholesta-5,8-dien-3β-ol
SMILES CCOC(=O)[N-]c1c[n+](no1)N2CCOCC2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21,23-24,28H,6-8,10-17H2,1-5H3/t19-,21+,23-,24+,26+,27-/m1/s1
Formula Weight 384.3392
Percent Purity >95%
Chemical Name or Material 8-Dehydrocholesterol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.