missing translation for 'onlineSavingsMsg'
Learn More

8-Deschloro-8-bromo Desloratadine, TRC

Product Code. 30138084
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138084 10 mg 10mg
30137643 5 mg 5mg
30003381 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138084 Supplier Toronto Research Chemicals Supplier No. TRCD29099510MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117796-50-6
Molecular Formula C19 H19 Br N2
Molecular Weight (g/mol) 355.27
Synonym Bromodesloratadine, 8-Bromo-6,11-dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
IUPAC Name 8-bromo-11-piperidin-4-ylidene-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMILES Brc1ccc2C(=C3CCNCC3)c4ncccc4CCc2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H19BrN2/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-22-19(14)18(17)13-7-10-21-11-8-13/h1-2,5-6,9,12,21H,3-4,7-8,10-11H2
Formula Weight 354.0732
Chemical Name or Material 8-Bromo-6,11-dihydro-11-(piperidin-4-ylidene)-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.