missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-2'-deoxyguanosine, MedChemExpress

Product Code. 16495447
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495447 50 mg 50mg
16484126 25 mg 25mg
16472678 5 mg 5mg
16400479 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495447 Supplier Medchem Express Supplier No. HYW011540
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 8-Hydroxy-2'-deoxyguanosine is a critical biomarker of oxidative stress and carcinogenesis.

8-Hydroxy-2'-deoxyguanosine is a critical biomarker of oxidative stress and carcinogenesis.

Specifications

Chemical Name or Material 8-Hydroxy-2'-deoxyguanosine
CAS 88847-89-6
Color White
Purity Grade Notes Research
Molecular Formula C10H13N5O5
Quantity 50 mg
Solubility Information DMSO : 155 mg/mL (547.24 mM; Need ultrasonic)
SMILES NC1=NC(C(NC2=O)=C(N2[C@H]3C[C@H](O)[C@H](O3)CO)N1)=O
Molecular Weight (g/mol) 283.24
Formula Weight 283.24
Percent Purity 96.17%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.