missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-2-methylquinoline, 98%

Product Code. 11387076
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387076 500 g 500g
11377076 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387076 Supplier Thermo Scientific Alfa Aesar Supplier No. L06989.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 826-81-3 | C10H9NO | 159.188 g/mol

8-Hydroxy-2-methylquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxy-2-methylquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 826-81-3
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00006765
InChI Key NBYLBWHHTUWMER-UHFFFAOYSA-N
Synonym 8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol
PubChem CID 13224
IUPAC Name 2-methylquinolin-8-ol
SMILES CC1=NC2=C(C=CC=C2O)C=C1

Specifications

Melting Point 70°C to 74°C
Boiling Point 267°C
Quantity 500 g
UN Number UN3077
Beilstein 119194
Solubility Information Insoluble in water.
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 8-Hydroxy-2-methylquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.