missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Quantikine Immunoassay Control Group 244 for Human IL-33
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
QC Control for ELISA Kits.
Specifications
Specifications
| For Use With (Application) | Control |
| Gene ID (Entrez) | 90865 |
| Host Species | Human |
| Quantity | 1 Ea. |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Target | IL-33 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction