missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy Mirtazapine, TRC

Product Code. 30135097
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135097 50 mg 50mg
30132549 25 mg 25mg
30007161 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135097 Supplier Toronto Research Chemicals Supplier No. TRCH94845050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102335-57-9
Molecular Formula C17H19N3O
Molecular Weight (g/mol) 281.35
Synonym Pyrazino[2,1-a]pyrido[2,3-c][2]benzazepin-8-ol, 1,2,3,4,10,14b-hexahydro-2-methyl-, 1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepin-8-ol, 8-Hydroxy-6-azamianserin, 8-Hydroxymirtazapine
SMILES CN1CCN2C(C1)c3ccccc3Cc4cc(O)cnc24

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H19N3O/c1-19-6-7-20-16(11-19)15-5-3-2-4-12(15)8-13-9-14(21)10-18-17(13)20/h2-5,9-10,16,21H,6-8,11H2,1H3
Formula Weight 281.1528
Chemical Name or Material 8-Hydroxy Mirtazapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.