missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyjulolidine, 97%

Product Code. 11433278
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433278 25 g 25g
11413278 1 g 1g
11423278 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433278 Supplier Thermo Scientific Alfa Aesar Supplier No. A17692.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 41175-50-2 | C12H15NO | 189.258 g/mol

8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41175-50-2
Molecular Formula C12H15NO
MDL Number MFCD00006918
InChI Key FOFUWJNBAQJABO-UHFFFAOYSA-N
PubChem CID 170474
IUPAC Name 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol

Specifications

Melting Point 132°C to 135°C
Quantity 25 g
Beilstein 1530423
Synonym 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1 h,5 h-benzo ij quinolizin-8-ol, 1 h,5 h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1 h,5 h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
Solubility Information Soluble aq. soln. (partly),ethanol,chloroform,methanol,and acetone.
SMILES OC1=CC=C2CCCN3CCCC1=C23
Molecular Weight (g/mol) 189.26
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 8-Hydroxyjulolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.