missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyjulolidine, 98%, ACROS Organics™

Product Code. 10421881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10421881 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10421881 Supplier Thermo Scientific Acros Supplier No. 296260010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 41175-50-2
CAS Min % 97.5
CAS Max % 100.0
Melting Point 132°C to 136°C
Color Beige to Pink or Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C12H15NO
MDL Number MFCD00006918
Quantity 1 g
Synonym 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
InChI Key FOFUWJNBAQJABO-UHFFFAOYSA-N
SMILES C1CC2=C3C(=C(C=C2)O)CCCN3C1
Molecular Weight (g/mol) 189.258
PubChem CID 170474
Formula Weight 189.26
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material 8-Hydroxyjulolidine, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.