missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline Copper Salt, TRC

Product Code. 30401932
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401932 5 g -
30400705 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401932 Supplier Toronto Research Chemicals Supplier No. TRCH9532685G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13014-03-4
Molecular Formula C9H7NO . x(Cu)
Molecular Weight (g/mol) 353.9
Synonym 8-Quinolinol Copper Salt
IUPAC Name copper;quinolin-8-olate
SMILES [Cu+2].[O-]c1cccc2cccnc12.[O-]c3cccc4cccnc34

Specifications

Quantity 5 g
InChI Formula InChI=1S/2C9H7NO.Cu/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;/h2*1-6,11H;/q;;+2/p-2
Formula Weight 351.0195
Chemical Name or Material 8-Hydroxyquinoline Copper Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.