missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline, TRC

Product Code. 17999939
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17999939 1 g 1g
30137751 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17999939 Supplier Toronto Research Chemicals Supplier No. TRCH9532651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 148-24-3
Formula Weight 145.0528
InChI Formula InChI=1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H
IUPAC Name quinolin-8-ol
Molecular Formula C9 H7 N O
Molecular Weight (g/mol) 145.16
SMILES Oc1cccc2cccnc12
Synonym 8-Quinolinol, 1-Azanaphthalene-8-ol, 8-Hydroxychinolin, 8-Hydroxyquinoline, 8-OQ, 8-Oxyquinoline, 8-Quinol, AQ+, Albisal, Fennosan H 30, NSC 2039, NSC 285166, NSC 402623, NSC 48037, NSC 54230, NSC 615011, NSC 82404, NSC 82405, NSC 82409, NSC 82410, NSC 82412, Oxin, Oxine, Oxoquinoline, Oxychinolin, Oxyquinoline, Phenopyridine, Quinophenol, Tumex
Chemical Name or Material 8-Hydroxyquinoline

Specifications

Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.