missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-d3 Psoralen, TRC

Product Code. 30477586
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477586 50 mg 50mg
30479401 25 mg 25mg
30465782 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477586 Supplier Toronto Research Chemicals Supplier No. TRCM26079850MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80386-99-8
Molecular Formula C12 2H3 H5 O4
Molecular Weight (g/mol) 219.21
Synonym 9-(Methoxy-d3)-7H-furo[3,2-g][1]benzopyran-7-one, Methoxsalen-d3
IUPAC Name 9-(trideuteriomethoxy)furo[3,2-g]chromen-7-one
SMILES [2H]C([2H])([2H])Oc1c2OC(=O)C=Cc2cc3ccoc13

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H8O4/c1-14-12-10-8(4-5-15-10)6-7-2-3-9(13)16-11(7)12/h2-6H,1H3/i1D3
Formula Weight 219.0611
Chemical Name or Material 8-Methoxy-d3 Psoralen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.