missing translation for 'onlineSavingsMsg'
Learn More

8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride, 90%, Thermo Scientific™

Product Code. 10314272
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10314272 1 g Amber Glass Bottle 1g
10374362 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314272 Supplier Thermo Scientific Maybridge Supplier No. BTB14320DA
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 646477-45-4
Molecular Formula C8H18Cl2N2
Molecular Weight (g/mol) 213.146
MDL Number MFCD07366481
InChI Key KVYQKWPZQKGICY-UHFFFAOYSA-N
Synonym 8-methyl-8-azabicyclo 3.2.1 octan-3-amine dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-amine dihydrochloride, 8-azabicyclo 3.2.1 octan-3-amine, 8-methyl-, dihydrochloride, 3-alpha-aminotropane dihydrochloride, 8-methyl-8-azabicyclo 3.2.1 oct-3-ylamine dihydrochloride, 3-amino-8-methyl-8-azabicyclo 3.2.1 octane dihydrochloride, 8-methyl-8-aza-bicyclo 3.2.1 octan-3-aminedihydrochloride
PubChem CID 2775928
IUPAC Name 8-methyl-8-azabicyclo[3.2.1]octan-3-amine;dihydrochloride
SMILES CN1C2CCC1CC(C2)N.Cl.Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.15
Percent Purity 90%
Chemical Name or Material 8-Methyl-8-azabicyclo[3.2.1]octan-3-amine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.