missing translation for 'onlineSavingsMsg'
Learn More

8-Nitroquinoline, 98%

Product Code. 11405607
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405607 5 g 5g
11415607 25 g 25g
11425607 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11405607 Supplier Thermo Scientific Alfa Aesar Supplier No. B23574.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 607-35-2 | C9H6N2O2 | 174.159 g/mol

8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Nitroquinoline is used in the preparation of bisubstituted phenoxy-6-methoxy-8-aminoquinoline. It is used to prepare furazano [3,4-h] quinoline.

Solubility
Soluble in ethanol, ethyl ether and benzene, slightly soluble in cold water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-35-2
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00006806
InChI Key OQHHSGRZCKGLCY-UHFFFAOYSA-N
Synonym quinoline, 8-nitro, unii-ywo6s88v4v, ccris 457, ywo6s88v4v, 8-nitroqunioline, 4msn, 8-nitro-quinoline, gilstone-yellow, zlchem 249, pubchem5825
PubChem CID 11830
IUPAC Name 8-nitroquinoline
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])N=CC=C2

Specifications

Melting Point 86°C to 90°C
Quantity 5 g
Beilstein 135178
Solubility Information Soluble in ethanol,ethyl ether and benzene,slightly soluble in cold water.
Formula Weight 174.16
Percent Purity 98%
Chemical Name or Material 8-Nitroquinoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.