missing translation for 'onlineSavingsMsg'
Learn More

8-Octanolactam, 98%

Product Code. 11428557
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428557 5 g 5g
11438557 25 g 25g
11448557 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11428557 Supplier Thermo Scientific Alfa Aesar Supplier No. A18621.06
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 935-30-8 | C8H15NO | 141.214 g/mol

2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).

Solubility
Soluble in methanol (almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 935-30-8
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.214
MDL Number MFCD00003273
InChI Key YDLSUFFXJYEVHW-UHFFFAOYSA-N
Synonym 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo
PubChem CID 13632
IUPAC Name azonan-2-one
SMILES C1CCCC(=O)NCCC1

Specifications

Melting Point 72°C to 75°C
Quantity 5 g
Beilstein 109775
Solubility Information Soluble in methanol (almost transparency).
Formula Weight 141.21
Percent Purity 98%
Chemical Name or Material 8-Octanolactam

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.