Learn More
Labconco 8-Port Manifold for CT-Series Freeze Dryer
Product Code. 31703096 Shop All Labconco Corporation Products
Change view
Click to view available options
Temperature Range (Metric):
-50°C
Unit Size:
Each
Product Code. Temperature Range (Metric) Unit Size
31703096 -50°C Each
1 options
Product Code. 31703096 Supplier Labconco Supplier No. 7232900
  • ONLINE EXCLUSIVE PRICE
    2323.32 €
    2012.74 € / Pack of 1
    Save 310.58 €
    13% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Manifold provides easy connection for adapters and flasks.
  • Manifold with eight neoprene valves with plastic knobs that accommodate both 1/2 in. and 3/4 in. adapters for connection of flask. For use with Labconco CT Series Freeze Dryers.
Specifications
Type Drying Manifold
No. of Shelves 0
Temperature Range (Metric) -50°C
Material Stainless Steel
Plug Type All except North America
Voltage 0/1 V