Learn More
Ibidi 8 Well Chamber, Removable, Bulk Pack, 8 per tray, 12 trays
Product Code. 17311339
Encompass
Change view
Click to view available options
Well Chamber
Product Code.
17311339 Well Chamber
1 options
Product Code. 17311339 Supplier Ibidi Supplier No. 8084196
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
8 Well Chamber, Removable, Bulk Pack, 8 per tray, 12 trays is a chamber slide with a removable 8 well silicone chamber.
  • Chamber slide with a removable 8 well silicone chamber for cell culture and immunofluorescence staining
  • for use with upright and inverted microscopes
  • bottom material: microscopy glass
  • packed in trays: 8 per tray
  • sterilized
Specifications
Type Chambered Coverglass
Wells 8
Sterility Sterile