Learn More
Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor
Support micro-volume protocols, such as PCR reaction set-up with this 8 x 8 PCR centrifuge rotor, capable of spinning up to 14,800rpm/ 16,800 x g.
Brand: Thermo Scientific™ 75005720
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Support micro-volume protocols, such as PCR reaction set-up, with the Thermo Scientific™ 8 x 8 PCR Strip Fixed Angle Rotor: Capable of spinning up to 15,000rpm / 17,860 x g for your PCR processing needs.
Supports micro-volume protocols, such as PCR reaction set-up, complete with our innovative Thermo Scientific technologies.
Secure, push-button Auto-Lock rotor exchange in less than 3 seconds delivers:
- Trouble-free rotor installation and removal
- No tools are required
- No complicated rotor changing procedures
- Clear chamber access for cleaning convenience
- Quick switch between applications, from conical tubes to blood tubes, from microtubes to microplates, and the flexibility to evolve with the changing needs of your laboratory.
Certified† biocontainment rotor lid for glove-friendly, one-handed operation.
Ordering Information:
Replacement biocontainment lid (75005730) and replacement O-rings (75005726) are sold separately. Contact your sales representative for more information.
Specifications
8 x 8 PCR Strip Fixed Angle Rotor | |
8 x 8 PCR Strips | |
15,000 rpm | |
553 | |
45° |
Fixed Angle Rotor | |
17,860 x g | |
71 mm | |
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges | |
Yes |
† Biocontainment certification by the Public Health England, Porton Down, UK.