missing translation for 'onlineSavingsMsg'
Learn More

80% Ethanol, Molecular Biology Grade

Product Code. 16649582 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2.5 L
4 L
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
2.5 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16649582 2.5 L Glass Bottle 2.5 L
16622284 4 L Poly Bottle 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 16649582 Supplier Thermo Scientific Supplier No. T08204DF2500
  • ONLINE EXCLUSIVE PRICE
    127.00 €
    123.00 € / 2.5 L
    Save 4.00 €
    3% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Ready made ethanol/water blend prequalified for molecular biology research applications. | CAS: 64-17-5 | C2H6O | 46.07 g/mol

  • 80% ethanol and water blend
  • DNase-, RNase-, and protease-free
  • Ready made ethanol/water blend
  • Pre-qualified for molecular biology research applications
  • Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
  • Tested for use with Applied Biosystems™ MagMAX™ kits
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 7732-18-5

Specifications

Color Clear, Colorless Liquid
Quantity 2.5 L
Packaging Glass Bottle
Residue after Evaporation 0.001% Maximum
Assay Percent Range 78% to 82%
Percent Purity 80%
Grade Molecular Biology
Chemical Name or Material Ethyl Alcohol
compliance-icons compliance-icons
Product Identifier
  • 80% Ethanol, for Molecular Biology, Duty Free
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol

For Manufacturing and Laboratory Use. Not for beverage use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.