missing translation for 'onlineSavingsMsg'
Learn More

80% Ethanol, for Water Molecular Biology, Thermo Scientific™

Product Code. 17535625 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17535625 Quantity: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 17535625 Supplier Thermo Scientific Supplier No. T082040500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Specifications

CAS 64-17-5, 7732-18-5
CAS Min % 74%, 26%
Color Colorless
CAS Max % 74%, 26%
Appearance Clear colorless liquid
Quantity 500 mL
Flash Point 21°C
Packaging Plastic
Physical Form Liquid
Residue after Evaporation 0.001 max.
Molecular Formula C2H6O
Linear Formula CH3CH2OH
MDL Number 00003568
Solubility Information Soluble in water
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Percent Purity 80%
Grade Molecular Biology
Chemical Name or Material 80% Ethanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.