Learn More
Hamilton™ 800 Microliter Syringes, No Needle Included
Product Code. 11530072
Change view
Click to view available options
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
11530072 Each
10001822 Each
11582355 Each
11572355 Each
11592355 Each
11550052 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11530072 Supplier Hamilton™ Supplier No. 764101
  • ONLINE EXCLUSIVE PRICE
    219.00 €
    179.44 € / Pack of 1
    Save 39.56 €
    18% Off
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 250 μL.
  • Ideal for dispensing volumes from 0.5 μL up to 250 μL
  • A variety of different terminations and needle options are available
  • Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending
Specifications
Material Borosilicate Glass
Needle Gauge No Needle Included
Product Type Microliter Syringe
For Use With (Equipment) Small Hub Removable Needle
Connection Type Removable
Graduated Yes