missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 800 TS Absorbance Reader
GREENER_CHOICE

Product Code. 17217799 Shop All Agilent BioTek Products
Click to view available options
Filters:
340, 405, 450, 490, 630 nm
405, 450, 490, 630 nm
Includes:
6- to 384-well Terasaki Plates
6- to 96-well Plates, Shaking
6- to 96-well Plates, Shaking, Incubation
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17217799

Brand: Agilent BioTek 800TSSN

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. For Research Use Only. Not for use in diagnostic procedures.

The BioTek 800 TS absorbance reader is an affordable, high-quality microplate reader for assays in 6- to 384-well formats. Applications include ELISA, protein quantification, enzyme kinetics and cell-based assays

The color touch screen provides a visual user interface, making programming fast and intuitive. Expanded functionality and data analysis is available with Gen5 software. The 800 TS partners with the 50 TS washer, making a compact, versatile, and affordable system to automate many application workflows.

Warranty

12 months

TRUSTED_SUSTAINABILITY

Especificaciones

Product Type Microplate Reader
Detector Type Photodiode
Interface USB Type B, USB Type A
Light Source Tungsten Halogen Lamp
Reading Options Endpoint, Kinetic, Well-area Scanning
Shaking Linear (except NB configurations)
Software Type Gen5 Data Analysis Software, Gen5 Secure for 21 CFR Part 11 compliance (option)
Bandwidth 10 nm
Certifications/Compliance CE and TUV marked. RoHS compliant.
Filters 405, 450, 490, 630 nm
For Use With (Application) Measuring absorbance in microplate-based assays
Includes 6- to 96-well Plates, Shaking
Model 800TS
Voltage 100/240 V
Wavelength Range 400 to 750 nm
Weight (English) 18.5 lb.
Weight (Metric) 8 kg
Mostrar más Mostrar menos
Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis