missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ 807 Manometric Module

Product Code. 16814520 Shop All Gilson Products
Change view
Click to view available options
Flow Rate:
10 to 200 mL/min.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate
16814520 10 to 200 mL/min.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16814520 Supplier Gilson™ Supplier No. 361737
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The 807 Manometric Module is designed for use with Gilson™ 305/306 piston-pumping systems. One manometric module is needed in each system to provide pulse-free operation and pressure monitoring.

  • Specially designed to meet the requirements of low pressure preparative liquid chromatography applications up to 8 MPa (80 bar or 1.2 kpsi)
  • Designed for use with Gilson™ 305/306 piston-pumping systems

Specifications

Flow Rate 10 to 200 mL/min.
Display Type Backlit LCD
Number of Channels 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.