missing translation for 'onlineSavingsMsg'
Learn More

Testo™ 830 T2 Infrared Thermometer

Carry out efficient, non-contact surface temperature measurements with this hygrometer. Testo™ 830 T2 Infrared Thermometer is ideal for undertaking control measurements in trade and industry.

Brand:  Testo™ 0560 8312

Product Code. 12692505

  • ONLINE EXCLUSIVE PRICE
    269.85 €
    102.00 € / Pack of 1
    Save 167.85 €
    62% Off
Estimated Shipment: 05-11-2026
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gun-style design is easy to handle
  • Integrated processor provides accurate measurement results
  • Infrared thermometer for non-contact surface temperature measurements
  • 12:1 optics and high resolution processor for accurate measurement results
  • Contact measurement possible with connectable temperature probe (TC type K)
  • Adjustable emission level
Specifications

Specifications

190 x 75 x 38 mm
0.1°C
-50°C to +500°C (TC Type K (NiCr-Ni)), 30°C to 400°C (Infrared)
ABS Plastic
200 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.