missing translation for 'onlineSavingsMsg'
Learn More

9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate, 97% (dry wt.), water ca 4-6%

Product Code. 11481527
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481527 2500 g 2500g
11461527 100 g 100g
11471527 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481527 Supplier Thermo Scientific Alfa Aesar Supplier No. B24969.0E
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 153277-35-1 | C14H7NaO5S | 310.255 g/mol

9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate. It is used as a catalyst in production of alkaline pulping in the soda process. It is used as an in situ photochemical fluorescence probe.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate forms charge-transfer complexes with 9,10-dimethoxyanthracene-2-sulfonate. It is used as a catalyst in production of alkaline pulping in the soda process. It is used as an in situ photochemical fluorescence probe.

Solubility
Soluble in hot water, cold water and insoluble in alcohol or ether.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153277-35-1
Molecular Formula C14H7NaO5S
Molecular Weight (g/mol) 310.255
MDL Number MFCD00149068
InChI Key GGCZERPQGJTIQP-UHFFFAOYSA-M
Synonym sodium anthraquinone-2-sulfonate, anthraquinone-2-sulfonic acid sodium salt, sodium 2-anthraquinonesulfonate, unii-511h72sdvx, sodium 9,10-dioxo-9,10-dihydroanthracene-2-sulfonate, 2-anthracenesulfonic acid, 9,10-dihydro-9,10-dioxo-, sodium salt, sodium 2-anthrachinonesulphonate, 2-anthraquinone sodium sulfonate, 2-anthraquinonesulfonic acid sodium salt, sodium-2-anthrachinonesulphonate
PubChem CID 23661981
IUPAC Name sodium;9,10-dioxoanthracene-2-sulfonate
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C=C(C=C3)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point >320°C
Quantity 2500 g
Beilstein 3580452
Solubility Information Soluble in hot water,cold water and insoluble in alcohol or ether.
Formula Weight 310.26 (Anhydrous)
Percent Purity 97%
Assay (dry wt.), water ≈4 to 6%
Chemical Name or Material 9,10-Anthraquinone-2-sulfonic acid sodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.