missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dioxo Ketotifen, TRC

Product Code. 30137792
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137792 50 mg 50mg
30007803 25 mg 25mg
30003962 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137792 Supplier Toronto Research Chemicals Supplier No. TRCD48651050MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 43076-16-0
Molecular Formula C19 H17 N O2 S
Molecular Weight (g/mol) 323.41
Synonym 4H-Benzo[4,5]cyclohepta[1,2-b]thiophene-9,10-dione, 4-(1-methyl-4-piperidinylidene)-, 4-(1-Methyl-4-piperidinylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophene-9,10-dione, NSC 282482, 4-(1-Methylpiperidin-4-ylidene)-4H-benzo[4,5]cyclohepta[1,2-b]thiophen-9,10-dione, 9,10-Dioxo Ketotifen, 9,10-Dioxoketotifen
IUPAC Name 10-(1-methylpiperidin-4-ylidene)benzo[1,2]cyclohepta[2,4-b]thiophene-4,5-dione
SMILES CN1CCC(=C2c3ccccc3C(=O)C(=O)c4sccc24)CC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C19H17NO2S/c1-20-9-6-12(7-10-20)16-13-4-2-3-5-14(13)17(21)18(22)19-15(16)8-11-23-19/h2-5,8,11H,6-7,9-10H2,1H3
Formula Weight 323.098
Percent Purity >95%
Chemical Name or Material 9,10-Dioxo Ketotifen
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.