missing translation for 'onlineSavingsMsg'
Learn More

9(10H)-Acridone, TRC

Product Code. 30134887
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134887 5 g 5g
17964145 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134887 Supplier Toronto Research Chemicals Supplier No. TRCA1909505G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 578-95-0
Molecular Formula C13 H9 N O
Molecular Weight (g/mol) 195.22
Synonym 9(10H)-Acridinone, 9-Acridanone (6CI,8CI), 9(10H)-Acridone, 9-Acridone, Acridanone, Acridin-9-one, Acridine, 9,10-dihydro-9-oxo-, Acridone, CK 103, CK 103 (heterocycle), NSC 408196, NSC 7664
IUPAC Name 10H-acridin-9-one
SMILES O=C1c2ccccc2Nc3ccccc13

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H9NO/c15-13-9-5-1-3-7-11(9)14-12-8-4-2-6-10(12)13/h1-8H,(H,14,15)
Formula Weight 195.0684
Percent Purity >95%
Chemical Name or Material Acridone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.