missing translation for 'onlineSavingsMsg'
Learn More

∆9(11)-Estradiol, TRC

Product Code. 30041284
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041284 10 mg 10mg
30041185 100 mg 100mg
30041266 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041284 Supplier Toronto Research Chemicals Supplier No. TRCE88803010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 791-69-5
Molecular Formula C18 H22 O2
Molecular Weight (g/mol) 270.37
Synonym Estra-1,3,5(10),9(11)-tetraene-3,17beta-diol, Estradiol Hemihydrate Imp. D (EP), 9,11-Didehydroestradiol, Estra-1,3,5(10),9(11)-tetraene-3,17-diol, (17β)-, Estra-1,3,5(10),9(11)-tetraene-3,17β-diol (6CI,7CI,8CI), (17β)-Estra-1,3,5(10),9(11)-tetraene-3,17-diol, 9,11-Dehydroestradiol, J 1213, ZYC 1, δ9,11-Estradiol
IUPAC Name (8S,13S,14S,17S)-13-methyl-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthrene-3,17-diol
SMILES C[C@]12CC=C3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@@H]2O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,8,10,15-17,19-20H,2,4,6-7,9H2,1H3/t15-,16+,17+,18+/m1/s1
Formula Weight 270.162
Percent Purity >95%
Chemical Name or Material 9,11-Didehydroestradiol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.