missing translation for 'onlineSavingsMsg'
Learn More

9-(4’-Hydroxyaminophenyl)-9H-pyrido[3,4-b]indole Hydrochloride, TRC

Product Code. 30405113
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405113 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30405113 Supplier Toronto Research Chemicals Supplier No. TRCH79500525MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-(4’-Hydroxyaminophenyl)-9H-pyrido[3,4-b]indole Hydrochloride
CAS 2512209-14-0
Molecular Formula C17H13N3O.HCl
Quantity 25 mg
Synonym Hydroxyaminophenylnorharman Hydrochloride, N-Hydroxy-4-(9H-pyrido[3,4-b]indol-9-yl)
InChI Formula InChI=1S/C17H13N3O.ClH/c21-19-12-5-7-13(8-6-12)20-16-4-2-1-3-14(16)15-9-10-18-11-17(15)20;/h1-11,19,21H;1H
SMILES ONC(C=C1)=CC=C1N2C3=C(C=CC=C3)C4=C2C=NC=C4.Cl
IUPAC Name N-(4-(9H-pyrido[3,4-b]indol-9-yl)phenyl)hydroxylamine hydrochloride
Molecular Weight (g/mol) 311.76
Formula Weight 311.57
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.