missing translation for 'onlineSavingsMsg'
Learn More

9-Aminoacridine hemihydrate, 98%

Product Code. 10596071 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10596071 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10596071 Supplier Thermo Scientific Acros Supplier No. 134410010
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 65944-23-2 | C13H10N2·1/2H2O | 203.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 65944-23-2, 90-45-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C to 235.0°C
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C13H10N2·1/2H2O
MDL Number MFCD00150520
Quantity 1 g
Beilstein 21, II, 280
Merck Index 14, 407
Synonym acridin-9-amine hydrate, 9-aminoacridine hemihydrate
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,freely soluble in alcohol,slightly soluble in chloroform,toluene and,pyridine
InChI Key SPWFQHCZHQCDAG-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N.O
IUPAC Name acridin-9-amine;hydrate
Molecular Weight (g/mol) 203.16
PubChem CID 2723763
Formula Weight 203.16
Percent Purity 98%
Chemical Name or Material 9-Aminoacridine hemihydrate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 9-Aminoacridine hemihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.