missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-Anthracenecarboxylic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 9-Anthracenecarboxylic Acid |
| CAS | 723-62-6 |
| Molecular Formula | C15H10O2 |
| Quantity | 10 g |
| Synonym | 9-Anthroic Acid, 9-Carboxyanthracene, NSC 151909, 9-Anthracenylcarboxylic Acid |
| InChI Formula | InChI=1S/C15H10O2/c16-15(17)14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-9H,(H,16,17) |
| SMILES | OC(=O)c1c2ccccc2cc3ccccc13 |
| IUPAC Name | anthracene-9-carboxylic acid |
| Molecular Weight (g/mol) | 222.24 |
| Formula Weight | 222.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction