missing translation for 'onlineSavingsMsg'
Learn More

9-Benzyl-6-chloro-7-deazapurine, 95%, Thermo Scientific™

Product Code. 15428897
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15428897

Brand: Thermo Scientific Alfa Aesar H52271.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
9-Benzyl-6-chloro-7-deazapurine is used as organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16019-34-4
Molecular Formula C13H10ClN3
Molecular Weight (g/mol) 243.694
MDL Number MFCD09953011
InChI Key KMWWCJXJJNHTQL-UHFFFAOYSA-N
Synonym 7-benzyl-4-chloro-7h-pyrrolo 2,3-d pyrimidine, 9-benzyl-6-chloro-7-deazapurine, amth011, 4-chloro-7-benzyl-7h-pyrrolo 2,3-d pyrimidine, 7-benzyl-4-chloro-7h-pyrrolo 2.3-d pyrimidine, 7-benzyl-4-chloropyrrolo 2,3-d pyrimidine
PubChem CID 15185356
IUPAC Name 7-benzyl-4-chloropyrrolo[2,3-d]pyrimidine
SMILES C1=CC=C(C=C1)CN2C=CC3=C2N=CN=C3Cl

Specifications

Quantity 1 g
Formula Weight 243.7
Percent Purity 95%
Chemical Name or Material 9-Benzyl-6-chloro-7-deazapurine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.