missing translation for 'onlineSavingsMsg'
Learn More

9-Bromo-9-phenylfluorene, TRC

Product Code. 30329112
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329112 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329112 Supplier Toronto Research Chemicals Supplier No. TRCB686455500MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Bromo-9-phenylfluorene
CAS 55135-66-5
Molecular Formula C19 H13 Br
Quantity 500 mg
InChI Formula InChI=1S/C19H13Br/c20-19(14-8-2-1-3-9-14)17-12-6-4-10-15(17)16-11-5-7-13-18(16)19/h1-13H
SMILES BrC1(c2ccccc2)c3ccccc3c4ccccc14
IUPAC Name 9-bromo-9-phenylfluorene
Molecular Weight (g/mol) 321.21
Formula Weight 320.0201
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.