missing translation for 'onlineSavingsMsg'
Learn More

9-Bromophenanthrene, 96%

Product Code. 10390494 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10390494 10 g Glass bottle 10g
10745941 25 g Glass bottle 25g
10776321 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10390494 Supplier Thermo Scientific Acros Supplier No. 107240100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 573-17-1 | C14H9Br | 257.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-17-1
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001174
InChI Key RSQXKVWKJVUZDG-UHFFFAOYSA-N
Synonym 9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene
PubChem CID 11309
IUPAC Name 9-bromophenanthrene
SMILES C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 60°C to 65°C
Color Yellow
Boiling Point 180°C to 190°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 05, 671
Formula Weight 257.13
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 9-Bromophenanthrene, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.