missing translation for 'onlineSavingsMsg'
Learn More

9-Bromophenanthrene, 98%

Product Code. 11332846
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332846 5 g 5g
11342846 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332846 Supplier Thermo Scientific Alfa Aesar Supplier No. L09033.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 573-17-1 | C14H9Br | 257.13 g/mol

9-Bromophenanthrene is used as halogenated building block. Isotope labelled 9-Bromophenanthrene (B687200), a halogenated polycyclic aromatic hydrocarbon that has been seen to have a room temperature phosphorescence that can be induced by β-cyclodextrin (β-CD) in the presence of cyclohexane; however, trace Fe(III) causes a decrease of the RTP emission.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Bromophenanthrene is used as halogenated building block. Isotope labelled 9-Bromophenanthrene (B687200), a halogenated polycyclic aromatic hydrocarbon that has been seen to have a room temperature phosphorescence that can be induced by β-cyclodextrin (β-CD) in the presence of cyclohexane; however, trace Fe(III) causes a decrease of the RTP emission.

Solubility
Soluble in toluene, and chloroform (50 mg/ml).

Notes
Incompatible with strong oxidizing agents. Store away from oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 573-17-1
Molecular Formula C14H9Br
Molecular Weight (g/mol) 257.13
MDL Number MFCD00001174
InChI Key RSQXKVWKJVUZDG-UHFFFAOYSA-N
Synonym 9-phenanthryl bromide, phenanthrene, 9-bromo, 9-phenathryl bromide, 9-bromo-phenanthrene, 9-bromo phenanthrene, 9-bromophenathrene, 9-bromanylphenanthrene, pubchem13993, acmc-1b0zz, 9-bromophenanthrene
PubChem CID 11309
IUPAC Name 9-bromophenanthrene
SMILES C1=CC=C2C(=C1)C=C(C3=CC=CC=C23)Br

Specifications

Melting Point 62°C to 66°C
Boiling Point 190°C (1.2 mmHg)
Quantity 5 g
Beilstein 1869927
Solubility Information Soluble in toluene,and chloroform (50mg/ml).
Formula Weight 257.14
Percent Purity 98%
Chemical Name or Material 9-Bromophenanthrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.