missing translation for 'onlineSavingsMsg'
Learn More

9-Chloro Triamcinolone Acetonide 21-Acetate, TRC

Product Code. 30350004
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350004 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350004 Supplier Toronto Research Chemicals Supplier No. TRCC4216851MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Chloro Triamcinolone Acetonide 21-Acetate
CAS 10392-75-3
Molecular Formula C26 H33 Cl O7
Quantity 1 mg
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-9-chloro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-chloro-11β,16α,17,21-tetrahydroxy-, cyclic 16,17-acetal with acetone, 21-acetate (7CI,8CI), 1H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-diene-3,20-dione deriv.
InChI Formula InChI=1S/C26H33ClO7/c1-14(28)32-13-20(31)26-21(33-22(2,3)34-26)11-18-17-7-6-15-10-16(29)8-9-23(15,4)25(17,27)19(30)12-24(18,26)5/h8-10,17-19,21,30H,6-7,11-13H2,1-5H3/t17-,18-,19-,21+,23-,24-,25-,26+/m0/s1
SMILES CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(Cl)[C@@H](O)C[C@]23C
IUPAC Name [2-[(1S,2S,4R,8S,9S,11S,12R,13S)-12-chloro-11-hydroxy-6,6,9,13-tetramethyl-16-oxo-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-14,17-dien-8-yl]-2-oxoethyl] acetate
Molecular Weight (g/mol) 492.989
Formula Weight 492.1915
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.