missing translation for 'onlineSavingsMsg'
Learn More

9’-Desmethyl Granisetron, TRC

Product Code. 30012430
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012430 50 mg 50mg
30010730 10 mg 10mg
30012726 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012430 Supplier Toronto Research Chemicals Supplier No. TRCD29204050MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 160177-67-3
Molecular Formula C17 H22 N4 O
Molecular Weight (g/mol) 298.38
Synonym Granisetron Hydrochloride Imp. C (EP), N-[(1R,3r,5S)-9-Azabicyclo[3.3.1]non-3-yl]-1-methyl-1H-indazole-3-carboxamide, N-(3-endo)-9-Azabicyclo[3.3.1]non-3-yl-1-methyl-1H-indazole-3-carboxamide, endo-N-9-Azabicyclo[3.3.1]non-3-yl-1-methyl-1H-indazole-3-carboxamide
IUPAC Name N-[(1R,5S)-9-azabicyclo[3.3.1]nonan-3-yl]-1-methylindazole-3-carboxamide
SMILES Cn1nc(C(=O)N[C@H]2C[C@H]3CCC[C@@H](C2)N3)c4ccccc14

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C17H22N4O/c1-21-15-8-3-2-7-14(15)16(20-21)17(22)19-13-9-11-5-4-6-12(10-13)18-11/h2-3,7-8,11-13,18H,4-6,9-10H2,1H3,(H,19,22)/t11-,12+,13+
Formula Weight 298.1794
Percent Purity >95%
Chemical Name or Material N-[(1R,3r,5S)-9-Azabicyclo[3.3.1]non-3-yl]-1-methyl-1H-indazole-3-carboxamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.