missing translation for 'onlineSavingsMsg'
Learn More

9-Fluorenone, TRC

Product Code. 30400680
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30400680 25 g 25g
30388879 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400680 Supplier Toronto Research Chemicals Supplier No. TRCF46278025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 9-Fluorenone
CAS 486-25-9
Molecular Formula C13 H8 O
Quantity 25 g
Synonym 9-Fluorenone, 9H-Fluoren-9-one, Fluoren-9-one (8CI), 9H-Fluorene-9-one, Fluorenone, NSC 5181
InChI Formula InChI=1S/C13H8O/c14-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8H
SMILES O=C1c2ccccc2c3ccccc13
IUPAC Name fluoren-9-one
Molecular Weight (g/mol) 180.2
Formula Weight 180.0575
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.