missing translation for 'onlineSavingsMsg'
Learn More

9-Formylphenanthrene, TRC

Product Code. 30397229
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397229 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397229 Supplier Toronto Research Chemicals Supplier No. TRCF7008851G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4707-71-5
Molecular Formula C15 H10 O
Quantity 1 g
Synonym Phenanthrene-9-aldehyde, 9-Phenanthrenecarboxaldehyde (6CI, 7CI, 8CI, 9CI, ACI), 9-Formylphenanthrene, 9-Phenanthraldehyde, 9-Phenanthrenaldehyde, 9-Phenanthrenecarbaldehyde, 9-Phenanthridinecarboxaldehyde, NSC 1932, Phenanthrene-9-carboxaldehyde
InChI Formula InChI=1S/C15H10O/c16-10-12-9-11-5-1-2-6-13(11)15-8-4-3-7-14(12)15/h1-10H
SMILES O=Cc1cc2ccccc2c3ccccc13
IUPAC Name phenanthrene-9-carbaldehyde
Molecular Weight (g/mol) 206.24
Formula Weight 206.0732
Chemical Name or Material Phenanthrene-9-aldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.